Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REFRACTIVE INDEX AAA LIQUID 1.395 1 * 1 Items
Catalog # CARG1803Y1.395
Supplier:
CARGILLE
Expand image
REFRACTIVE INDEX AAA LIQUID 1.395 1 * 1 Items
Catalog # CARG1803Y1.395
Supplier:
CARGILLE