0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, both SMMMS 50g/m², one blue, one green, 120x120cm, DIN EN ISO 116 07 1 * 50 items
Catalog # VEPA3FDOP383218
Supplier:
VP Medical Packaging
Expand image
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, both SMMMS 50g/m², one blue, one green, 120x120cm, DIN EN ISO 116 07 1 * 50 items
Catalog # VEPA3FDOP383218
Supplier:
VP Medical Packaging