0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 300.8 MPA.S (25.00C), RTM18 1 * 500 mL
Catalog # PARGRTM18
Supplier:
PARAGON SCIENTIFIC
Expand image
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 300.8 MPA.S (25.00C), RTM18 1 * 500 mL
Catalog # PARGRTM18
Supplier:
PARAGON SCIENTIFIC