Order Entry
Norway
ContactUsLinkComponent
Detect Mouse Mcp1/CCL2 with <6pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supe rnates and serum. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. 1 * 1 KIT
Catalog # BSBTFEK0568
Supplier:  Boster Bio
Detect Mouse Mcp1/CCL2 with <6pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supe rnates and serum. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. 1 * 1 KIT
Catalog # BSBTFEK0568
Supplier:  Boster Bio
Frequently Bought Together