Order Entry
Norway
ContactUsLinkComponent
Detect Mouse BMP2 with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: bone tissue, cell cult ure supernates and serum. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set 1 * 1 KIT
Catalog # BSBTFEK0313
Leverandør:  Boster Bio
Detect Mouse BMP2 with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: bone tissue, cell cult ure supernates and serum. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set 1 * 1 KIT
Catalog # BSBTFEK0313
Leverandør:  Boster Bio
Frequently Bought Together