Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STORAGE BOX HALF MASK 1 * 1 items
Catalog # MOLD799501
Supplier:
Moldex
Utvid bildet
STORAGE BOX HALF MASK 1 * 1 items
Catalog # MOLD799501
Supplier:
Moldex