0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TG-PBDE GC COLUMN, 15M X 0.25MM X 0.10UM 1 * 1 items
Catalog # HYPE26061-0350
Supplier:
Thermo Fisher Scientific
Expand image
TG-PBDE GC COLUMN, 15M X 0.25MM X 0.10UM 1 * 1 items
Catalog # HYPE26061-0350
Supplier:
Thermo Fisher Scientific