Order Entry
Norway
ContactUsLinkComponent
A Stc1 (Stanniocalcin-1) ELISA with a senstivity of 46.9pg/ml and a range of 78-5000pg/ml. Uses the detection method Sandwich ELISA, Double Antibody 1 * 1 KIT
  GENOIT8378
A Stc1 (Stanniocalcin-1) ELISA with a senstivity of 46.9pg/ml and a range of 78-5000pg/ml. Uses the detection method Sandwich ELISA, Double Antibody 1 * 1 KIT
  GENOIT8378
Frequently Bought Together