Order Entry
Norway
ContactUsLinkComponent
A BCKDK ELISA with a senstivity of 46.9pg/ml and a range of 78-5000pg/ml. Uses the detection method Sandwich ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8364
Supplier:  G-Biosciences
A BCKDK ELISA with a senstivity of 46.9pg/ml and a range of 78-5000pg/ml. Uses the detection method Sandwich ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8364
Supplier:  G-Biosciences
Frequently Bought Together