Order Entry
Norway
ContactUsLinkComponent
An ELISA kit for the detection of MMP-13 (Rat). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.188ng/ml. 1 * 1 KIT
Catalog # GENOIT7504
Supplier:  G-Biosciences
An ELISA kit for the detection of MMP-13 (Rat). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.188ng/ml. 1 * 1 KIT
Catalog # GENOIT7504
Supplier:  G-Biosciences
Frequently Bought Together