Order Entry
Norway
ContactUsLinkComponent
An ELISA kit for the detection of MMP-12 (Rat). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 46.875pg/ml. 1 * 1 KIT
Catalog # GENOIT7503
Supplier:  G-Biosciences
An ELISA kit for the detection of MMP-12 (Rat). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 46.875pg/ml. 1 * 1 KIT
Catalog # GENOIT7503
Supplier:  G-Biosciences
Frequently Bought Together