kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
An ELISA kit for the detection of MMP-10 (Mouse). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 37.5pg/ml. 1 * 1 KIT
GENOIT5042
:
G-Biosciences
An ELISA kit for the detection of MMP-10 (Mouse). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 37.5pg/ml. 1 * 1 KIT
GENOIT5042
:
G-Biosciences