Order Entry
Norway
ContactUsLinkComponent
An ELISA kit for the detection of Pro-MMP-13 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 46.875pg/ml . 1 * 1 KIT
Catalog # GENOIT4291
Supplier:  G-Biosciences
An ELISA kit for the detection of Pro-MMP-13 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 46.875pg/ml . 1 * 1 KIT
Catalog # GENOIT4291
Supplier:  G-Biosciences
Frequently Bought Together