Order Entry
Norway
ContactUsLinkComponent
An ELISA kit for the detection of Pro-MMP-10 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 46.875pg/ml . 1 * 1 KIT
  GENOIT4290
An ELISA kit for the detection of Pro-MMP-10 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 46.875pg/ml . 1 * 1 KIT
  GENOIT4290
Frequently Bought Together