Order Entry
Norway
ContactUsLinkComponent
An ELISA kit for the detection of MMP-25 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.094ng/ml. 1 * 1 KIT
Katalog # GENOIT4135
Supplier:  G-Biosciences
An ELISA kit for the detection of MMP-25 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.094ng/ml. 1 * 1 KIT
Katalog # GENOIT4135
Supplier:  G-Biosciences