Order Entry
Norway
ContactUsLinkComponent
An ELISA kit for the detection of MMP-12 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.188ng/ml. 1 * 1 KIT
Katalog # GENOIT1686
Supplier:  G-Biosciences
An ELISA kit for the detection of MMP-12 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.188ng/ml. 1 * 1 KIT
Katalog # GENOIT1686
Supplier:  G-Biosciences