0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Stericlin® sterilisation wrap sheet, medical paper crepe, white, 100x100cm, DIN EN ISO 11607 1 * 250 items
Catalog # 129-1441
Supplier:
VP Medical Packaging
Utvid bildet
Stericlin® sterilisation wrap sheet, medical paper crepe, white, 100x100cm, DIN EN ISO 11607 1 * 250 items
Catalog # 129-1441
Supplier:
VP Medical Packaging