kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Stericlin® sterilisation wrap sheet, non-woven 59g/m² blue, 75x75cm, DIN EN ISO 11607 1 * 200 items
129-1389
:
VP Medical Packaging
Stericlin® sterilisation wrap sheet, non-woven 59g/m² blue, 75x75cm, DIN EN ISO 11607 1 * 200 items
129-1389
:
VP Medical Packaging