5-Hydroxyisoindolin-1-one
Bottle, 1 g
Catalog # APOSOR305282-1G
Supplier: Apollo Scientific
5-Hydroxyisoindolin-1-one
Catalog # APOSOR305282-1G
Supplier: Apollo Scientific
CAS Number: 252061-66-8
Jump to:
Product Details & Documents
- CAS Number:252061-66-8
- Formula:C8H7NO2
- MDL Number:MFCD10000830
- Molecular Weight:149.15 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



