Order Entry
Norway
Orders LinkContactUsLinkComponent
CapLock™ clips
CapLock™ clips
CapLock™ clip, Red
Catalog # SIMPT330-75CLR
Supplier:   Simport Scientific
CapLock™ clips
Catalog # SIMPT330-75CLR
Supplier:  Simport Scientific
CapLock™ clips
CapLock™ clips
CapLock™ clip, Red
Price
kr 455,00
Log in for availability
1 * 100 Items
Product Details & Documents
These CapLock™ clips are made of PE. They provide extra security against inadvertent cap openings on critical samples or during incubation above 80 °C.
  • Ideal for shipping samples
  • Suitable for 5 ml tubes with snap cap
Specifications
  • Colour:
    Red
  • Description:
    CapLock™ clip
  • Seq:
    67