0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Rotor seal 6 port 2-pos C2/C3-valve1/16*.75mm, 75C/4000psi liq,
Catalog # VICIVV-C2-30R6
Supplier:
VICI
Expand image
Rotor seal 6 port 2-pos C2/C3-valve1/16*.75mm, 75C/4000psi liq,
Catalog # VICIVV-C2-30R6
Supplier:
VICI