Order Entry
Norway
ContactUsLinkComponent
Hanks' Balanced Salt Solution (1X), without calcium, without magnesium, without phenol red 1 * 500 mL
  J67681.AP
Hanks' Balanced Salt Solution (1X), without calcium, without magnesium, without phenol red 1 * 500 mL
  J67681.AP
Frequently Bought Together