4'-Bromoacetophenone
Bottle, 100 g
APOSOR01684-100G
: p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
- :99-90-1
- :C₈H₇BrO
- :MFCD00000105
- :199.05 g/mol
- :255…256 °C
- :49…53 °C
- :1.647 g/cm³ (25 °C)
- :>110 °C
- :Ambient
- :202-799-3
- Pack type:Bottle
- Pk:100 g