5-Nitroisoindolin-1-one
Bottle, 1 g
Catalog # APOSOR305280-1G
Supplier: Apollo Scientific
5-Nitroisoindolin-1-one
Catalog # APOSOR305280-1G
Supplier: Apollo Scientific
CAS Number: 876343-38-3
Gå til:
Product Details & Documents
Synonymer: 2,3-Dihydro-5-nitro-1H-isoindol-1-one, 2,3-Dihydro-5-nitro-1-oxo-1H-isoindole
- CAS Number:876343-38-3
- Formel:C₈H₆N₂O₃
- MDL-nummer:MFCD10000827
- Molecular Weight:178.145 g/mol
- Lagringstemperatur:Ambient
Spesifikasjoner
Spesifikasjoner
- Pack type:Bottle
- Pk:1 g
Spesifikasjoner
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



