Morpholin-3-one
Bottle, 5 g
Catalog # APOSOR301177-5G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 3-Morpholinone
- CAS Number:109-11-5
- Formula:C₄H₇NO₂
- MDL Number:MFCD00631009
- Molecular Weight:101.1039 g/mol
- Melting Pt:104…107 °C
- Storage Temperature:Ambient
- EINECS:203-647-9
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g



