Morpholin-3-one
Bottle, 100 g
Catalog # APOSOR301177-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 3-Morpholinone
- CAS Number:109-11-5
- Formula:C₄H₇NO₂
- MDL Number:MFCD00631009
- Molecular Weight:101.1039 g/mol
- Melting Pt:104…107 °C
- Storage Temperature:Ambient
- EINECS:203-647-9
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



