Order Entry
Norway
Orders LinkContactUsLinkComponent
2-Phenylpropionic acid 97%

2-Phenylpropionic acid 97%

Bottle, 5 g
Catalog # APOSOR2976-5G
Irritant
Supplier:   Apollo Scientific
2-Phenylpropionic acid 97%
Catalog # APOSOR2976-5G
Supplier:  Apollo Scientific
CAS Number:  492-37-5
2-Phenylpropionic acid 97%

Bottle, 5 g

CAS Number:  492-37-5
Price
kr270.00
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid
  • CAS Number:492-37-5
  • Formula:C₉H₁₀O₂
  • MDL Number:MFCD00002650
  • Molecular Weight:150.18 g/mol
  • Boiling Pt:260…266 °C
  • Melting Pt:–5 °C
  • Density:1.1 g/cm³ (25 °C)
  • Flash Pt:110 °C
  • Storage Temperature:Ambient
  • EINECS:207-752-0
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.