3-Methoxyoxan-4-one
Bottle, 1 g
Catalog # APOSOR55049-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 3-Methoxy-tetrahydropyran-4-one, 3-Methoxydihydro-2H-pyran-4(3H)-one
- CAS Number:624734-17-4
- Formula:C₆H₁₀O₃
- MDL Number:MFCD09909903
- Molecular Weight:130.142 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g
- Cat. No.:APOSOR55049-1G



