Boc-Pro-OH
Bottle, 25 g
Catalog # APOSOR61282-25G
Supplier: Apollo Scientific
Gå til:
Product Details & Documents
Synonymer: N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid
- CAS Number:15761-39-4
- Formel:C₁₀H₁₇NO₄
- MDL-nummer:MFCD00037324
- Molecular Weight:215.25 g/mol
- Molten Pt:133…136 °C
- Lagringstemperatur:Refrigerator
- EINECS:239-848-3
Spesifikasjoner
Spesifikasjoner
- Pack type:Bottle
- Pk:25 g
Spesifikasjoner
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



