Boc-Pro-OH
Bottle, 100 g
Catalog # APOSOR61282-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid
- CAS Number:15761-39-4
- Formula:C₁₀H₁₇NO₄
- MDL Number:MFCD00037324
- Molecular Weight:215.25 g/mol
- Melting Pt:133…136 °C
- Storage Temperature:Refrigerator
- EINECS:239-848-3
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g



