Cefaclor
Bottle, 1 g
Catalog # APOSBIM0109-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: (6R,7R)-7-((R)-2-Amino-2-phenylacetamido)-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
- CAS Number:53994-73-3
- Formula:C₁₅H₁₄ClN₃O₄S
- MDL Number:MFCD00151471
- Molecular Weight:385.8 g/mol
- Storage Temperature:Ambient
- EINECS:258-909-5
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



