Order Entry
Norway
Orders LinkContactUsLinkComponent
ADA ≥98%

ADA ≥98%

Bottle, 100 g
Catalog # APOSBIA0302-100G
Irritant
Supplier:   Apollo Scientific
ADA ≥98%
Catalog # APOSBIA0302-100G
Supplier:  Apollo Scientific
CAS Number:  26239-55-4
ADA ≥98%

Bottle, 100 g

CAS Number:  26239-55-4
Price
kr 305,00
Log in for availability
1 * 100 g
Product Details & Documents

Biological buffer - useful range 6.0 - 7.2

Synonyms:  2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid
  • CAS Number:26239-55-4
  • Formula:H₂NC(O)CH₂N(CH₂COOH)₂
  • MDL Number:MFCD00008031
  • Molecular Weight:190.16 g/mol
  • Melting Pt:219 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications