Order Entry
Norway
Orders LinkContactUsLinkComponent
Capillary tubes

Capillary tubes

100 mm, 1,5 mm, 0,75
Catalog # HILG1400372
Supplier:   HILGENBERG
Capillary tubes
Catalog # HILG1400372
Supplier:  HILGENBERG
Capillary tubes
Capillary tubes

100 mm, 1,5 mm, 0,75

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

Borosilicate glass, ends cut, with filament.

Specifications
  • Ext.Ø:
    1,5 mm
  • Ø int.:
    0,75
  • Length:
    100 mm
  • Seq:
    09
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.