Cefaclor 96%
Glass bottle, 5 g
Catalog # ACRO455260050
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (6R,7R)-7-((R)-2-Amino-2-phenylacetamido)-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
- CAS Number:53994-73-3
- Formula:C₁₅H₁₄ClN₃O₄S
- MDL Number:MFCD00151471
- Molecular Weight:385.8 g/mol
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Water:3.0 to 9.0 %
- HPLC:95.0 %
Specifications
- Pack type:Glass bottle
- Pk:5 g



