Morpholin-3-one 98%
Glass bottle, 25 g
Catalog # ACRO460530250
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 3-Morpholinone
- CAS Number:109-11-5
- Formula:C₄H₇NO₂
- MDL Number:MFCD00631009
- Molecular Weight:101.1039 g/mol
- Melting Pt:106…108 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:Colorless to white Crystalline solid
- Infrared spectrum:Conforms
- Melting point:102 to 109 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:25 g



