Morpholin-3-one 98%
Glass bottle, 1 g
Catalog # ACRO460530010
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 3-Morpholinone
- CAS Number:109-11-5
- Formula:C₄H₇NO₂
- MDL Number:MFCD00631009
- Molecular Weight:101.1039 g/mol
- Melting Pt:106…108 °C
- Storage Temperature:Romtemperatur
Specifications
Specification Test Results
- Appearance:Colorless to white Crystalline solid
- Infrared spectrum:Conforms
- Melting point:102 to 109 °C
- GC:≥97.5 %
Specifications
- Pakketype:Glass bottle
- Fp:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



