KCM 1-B from Behr Labor-Technik Kompakt system with magnetic stirrer for liberation and separation operations in the determination of total cyanide in soil samples (40 ml absorption vessel) 1 * 1 items
Katalog # BEHRB00217735
Supplier: BEHR
Frequently Bought Together