Order Entry
Norway
ContactUsLinkComponent
This gene encodes a protein which contains a hinge region domain found in members of the SMC (structural maintenance of chromosomes) protein family. 1 * 100 µG
Catalog # BWRLBS8781
This gene encodes a protein which contains a hinge region domain found in members of the SMC (structural maintenance of chromosomes) protein family. 1 * 100 µG
Catalog # BWRLBS8781
Frequently Bought Together