The protein encoded by this gene is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisti ng of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzy 1 * 100 µG
Catalog # BWRLBS8607
Supplier: Bioworld Technology
Frequently Bought Together