0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
4 – 20 ma current loop interface, o2 controller, actual value (0 – 25 % o2 = 4 – 20 ma) 1 * 1 items
Catalog # MEMMICO150V1
Supplier:
MEMMERT
Expand image
4 – 20 ma current loop interface, o2 controller, actual value (0 – 25 % o2 = 4 – 20 ma) 1 * 1 items
Catalog # MEMMICO150V1
Supplier:
MEMMERT