Order Entry
Norway
ContactUsLinkComponent
Control of oxygen concentration by n2 inlet adjustment range 1 % up to 20 % o2 setting accuracy 0.1 %. (requires option k7) 1 * 1 items
Catalog # MEMMICO105T6
Supplier:  MEMMERT
Control of oxygen concentration by n2 inlet adjustment range 1 % up to 20 % o2 setting accuracy 0.1 %. (requires option k7) 1 * 1 items
Catalog # MEMMICO105T6
Supplier:  MEMMERT