Order Entry
Norway
ContactUsLinkComponent
G-Sep Agarose 6B is a gel filtration matrix formed from agarose beads that is available with 6% agarose content, designated G-Sep Agarose 6B. The resin is a proven gel filtration base matrix and is routinely modified by researchers to couple affini 1 * 1 L
Catalog # GENO786-955
Supplier:  G-Biosciences
G-Sep Agarose 6B is a gel filtration matrix formed from agarose beads that is available with 6% agarose content, designated G-Sep Agarose 6B. The resin is a proven gel filtration base matrix and is routinely modified by researchers to couple affini 1 * 1 L
Catalog # GENO786-955
Supplier:  G-Biosciences
Related Information