Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REFRACTIVE INDEX STD SERIE E RI=1,640 1 * 7,4 mL
Catalog # CARG1843X1.640
Supplier:
CARGILLE
Expand image
REFRACTIVE INDEX STD SERIE E RI=1,640 1 * 7,4 mL
Catalog # CARG1843X1.640
Supplier:
CARGILLE