Order Entry
Norway
Orders LinkContactUsLinkComponent
pH buffer solutions

pH buffer solutions

pH buffer standard solution with certificate of analysis, 250 ml, 4,01, 250 mL
Catalog # HACHLZW9463.99
Supplier:   Hach
pH buffer solutions
Catalog # HACHLZW9463.99
Supplier:  Hach
CAS Number:  
pH buffer solutions
pH buffer solutions

pH buffer standard solution with certificate of analysis, 250 ml, 4,01, 250 mL

CAS Number:  
Price
kr 300,00
Log in for availability
1 * 250 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    pH buffer standard solution with certificate of analysis, 250 ml
  • pH value:
    4,01

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.