kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
STORAGE BIN SIZE 5 GREEN 1 * 1 items
Catalog # HUEN675400
Supplier:
hünersdorff GmbH
Expand image
STORAGE BIN SIZE 5 GREEN 1 * 1 items
Catalog # HUEN675400
Supplier:
hünersdorff GmbH