TETRADECYLTRIMETHYL-AMMONIUM BROMIDE. (Myristyltrimethylammonium bromide) Purity: 98% Phase transfer catalyst. 1 * 100 g
ICNA0215210980
TETRADECYLTRIMETHYL-AMMONIUM BROMIDE. (Myristyltrimethylammonium bromide) Purity: 98% Phase transfer catalyst. 1 * 100 g
ICNA0215210980
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection