Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REPLACEMENT HYGROMETER. Replacement hygrometer for Dry 10 desiccator. 1 * 1 items
Catalog # ICNA0219949801
Supplier:
MP Biomedicals
Expand image
REPLACEMENT HYGROMETER. Replacement hygrometer for Dry 10 desiccator. 1 * 1 items
Catalog # ICNA0219949801
Supplier:
MP Biomedicals