Order Entry
Norway
ContactUsLinkComponent
An -6 isomer of docosapentaenoic acid formed by the elongation and desaturation of arachidonic acid 1 * 5 mg
Catalog # CAYM10008335-5
Supplier:  Cayman Chemical
An -6 isomer of docosapentaenoic acid formed by the elongation and desaturation of arachidonic acid 1 * 5 mg
Catalog # CAYM10008335-5
Supplier:  Cayman Chemical