Order Entry
Norway
ContactUsLinkComponent
A potent allosteric inhibitor of PRMT3 (IC50 = 50 nM) with >100-fold selectivity over other methyltransferases and other non-epigene tic targets 1 * 25 mg
  CAYM17017-25
A potent allosteric inhibitor of PRMT3 (IC50 = 50 nM) with >100-fold selectivity over other methyltransferases and other non-epigene tic targets 1 * 25 mg
  CAYM17017-25
Frequently Bought Together