Order Entry
Norway
ContactUsLinkComponent
TRESPA TopLabPlus worksurface with retention tray SMART483. Ideal for powders handlings. Weighing: precision up to 6 decimal places. High ch emical and mecanical resistance. Easy cleaning 1 * 1 items
Catalog # ERLAI0164030001
Supplier:  ERLAB CAPTAIRE
TRESPA TopLabPlus worksurface with retention tray SMART483. Ideal for powders handlings. Weighing: precision up to 6 decimal places. High ch emical and mecanical resistance. Easy cleaning 1 * 1 items
Catalog # ERLAI0164030001
Supplier:  ERLAB CAPTAIRE
Frequently Bought Together